ContactUsLinkComponent
SONOREX TECHNIK RM 112 UH, 40 kHz, ultrasonic bath, 400V 3N~ (± 10%) 50/60 Hzz, ID 600x450x450/470 mm with inclined tank bottom 145 l, welded tank 2 mm s/s AISI 316 Ti timer 1-15 min and continuous, heating 30-80°C, ultrasonic peak output 4000 W, H 1 * 1 items
Catalog # BAND9102
undefined
SONOREX TECHNIK RM 112 UH, 40 kHz, ultrasonic bath, 400V 3N~ (± 10%) 50/60 Hzz, ID 600x450x450/470 mm with inclined tank bottom 145 l, welded tank 2 mm s/s AISI 316 Ti timer 1-15 min and continuous, heating 30-80°C, ultrasonic peak output 4000 W, H 1 * 1 items
Catalog # BAND9102
Frequently Bought Together