Vitamin D ELISA kit
Product Details & Documents
Sensitive ELISA for vitamin D quantification in plasma or serum.
- Measure vitamin D in plasma or serum
- Detect the metabolically stable forms 25(OH)D₃ and 25(OH)D₂
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure vitamin D levels down to 0,5 ng/ml
- Read: Colorimetric at 405 to 420 nm
The primary synthesis of functional vitamin D begins in the skin, where a cholesterol by-product acted upon by UV light forms vitamin D₃. Vitamin D₃ is carried through the bloodstream, attached to the vitamin D binding protein, to the liver where it is converted to 25-hydroxy vitamin D₃ (25(OH)D₃). This product is then transported to the kidney where it is further converted to 1,25-dihydroxycholecalciferol (1,25(OH)₂D₃), the biologically active form. Active 1,25(OH)₂D₃ is very short-lived and is rapidly metabolized to its deactivated forms.
Vitamin D Immunoassay, accurate Vitamin D measurement in plasma or serum. Vitamin D ELISA Kit is a competitive assay that can be used for Vitamin D quantification in plasma or serum. Because of the short half-life of the biologically active 1,25(OH)D form, this Vitamin D assay primarily detects the more metabolically stable forms, 25(OH)D₃ and 25(OH)D₂.
Accurate detection of these forms requires that they be displaced from the vitamin D binding protein prior to measurement. A simple, acetone-based purification protocol is included as the initial procedure of the Vitamin D assay kit for this purpose.
Packaging: Kit contains: Antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, tracer and antiserum dye.
Documents
TDS Vitamin D ELISA Kit
Datasheet
Specifikace
Specifikace
- Pk:96 Tests
- Description:Vitamin D ELISA Kit, 96 strip wells
- Cat. No.:CAYM501050-96
- Mid point:1 - 4 ng/ml (50% B/B₀)
- Developmental time:60 - 90 min
- Sensitivity:0,5 ng/ml (80% B/B₀)
- Detection range:0,19 - 25 ng/ml
- Storage temperature:–20 °C
- Incubation time:18 hr
Specifikace
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



