ContactUsLinkComponent
Tubing for pressure-sided connection to the microsart e.jet transfer pump the required length depends on the distance between transfer pump and drain. length: 10 m. 1 * 1 items
Catalog # 512-0738
Supplier:  Sartorius
Tubing for pressure-sided connection to the microsart e.jet transfer pump the required length depends on the distance between transfer pump and drain. length: 10 m. 1 * 1 items
Catalog # 512-0738
Supplier:  Sartorius
Frequently Bought Together