Viscosity Standards(cSt) reads 320@20C, 230@25C, [email protected], 98@40C, 61@50C 1 * 500 mL
Catalog # REGEREVIS-N100
Supplier: REAGECON
Viscosity Standards(cSt) reads 320@20C, 230@25C, [email protected], 98@40C, 61@50C 1 * 500 mL
Catalog # REGEREVIS-N100
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW