ContactUsLinkComponent
DNAmark™ 20bp Ladder consists of 13 fragments ranging from 60-300 base pairs (bp). This molecular weight marker is specially designe d for identifying small DNA fragments and allowing for quantitative estimation of the DNA fragments in the gel at the 1 * 50 µG
Catalog # GENO786-852
Supplier:  G-Biosciences
DNAmark™ 20bp Ladder consists of 13 fragments ranging from 60-300 base pairs (bp). This molecular weight marker is specially designe d for identifying small DNA fragments and allowing for quantitative estimation of the DNA fragments in the gel at the 1 * 50 µG
Catalog # GENO786-852
Supplier:  G-Biosciences