ContactUsLinkComponent
Sodium metaperiodate, or sodium m-periodate, is a mild oxidant that is routinely used for the conversion of cis-glycol groups in car bohydrates to reactive aldehdye groups. The reactive aldehyde groups are used in chemical conjugation procedures or r 1 * 25 g
Catalog # GENOBKC-12
Supplier:  G-Biosciences
Sodium metaperiodate, or sodium m-periodate, is a mild oxidant that is routinely used for the conversion of cis-glycol groups in car bohydrates to reactive aldehdye groups. The reactive aldehyde groups are used in chemical conjugation procedures or r 1 * 25 g
Catalog # GENOBKC-12
Supplier:  G-Biosciences