ContactUsLinkComponent
Bialaphos is a tripeptide antibiotic that is used in transformation experiments of many species of plants that make use of the bar gene for selection. In corn it is more effective than glufosinate. In wheat, bialaphos was the most reliable selection 1 * 0,1 g
Catalog # GENORC-158
Supplier:  G-Biosciences
Bialaphos is a tripeptide antibiotic that is used in transformation experiments of many species of plants that make use of the bar gene for selection. In corn it is more effective than glufosinate. In wheat, bialaphos was the most reliable selection 1 * 0,1 g
Catalog # GENORC-158
Supplier:  G-Biosciences
Frequently Bought Together