Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine free base

3,3',5,5'-Tetramethylbenzidine free base

Bottle
Catalog # GENORC-317
Supplier:   G-Biosciences
3,3',5,5'-Tetramethylbenzidine free base
Catalog # GENORC-317
Supplier:  G-Biosciences
CAS Number:  
3,3',5,5'-Tetramethylbenzidine free base

Bottle

CAS Number:  
Price
23 740,00 Kč
Log in for availability
1 * 50 g
Product Details & Documents

TMB is a widely used chromogenic substrate for horseradish peroxidase (HRP) in colorimetric assays. As a free base, it is typically dissolved in an organic solvent or buffer to prepare TMB substrate solutions for ELISA, Western blotting, and immunohistochemistry.

  • Sensitive HRP Substrate: Enables high-sensitivity detection in enzyme-linked assays
  • Colorimetric Detection: Produces a strong blue signal, with a yellow endpoint upon acid stop for dual-wavelength measurement
  • Flexible Formulation: Free base form allows custom preparation for specific assay conditions
  • Versatile Applications: Ideal for ELISA, Western blotting, and immunoassays in diagnostics and research
  • Low Background: Provides high signal-to-noise ratio for accurate quantification
Specifications

Specifications

  • Pack type:
    Bottle

Specifications