ContactUsLinkComponent
Gentamicin efficiently inhibits growth of many Gram-positive and Gram-negative microorganisms. It does so by inhibiting protein synt hesis by binding to the L6 protein of the 30S ribosomal subunit. It is also effective against mycoplasma. Recommended 1 * 25 g
Katalogové číslo GENORC-175
Supplier:  G-Biosciences
Gentamicin efficiently inhibits growth of many Gram-positive and Gram-negative microorganisms. It does so by inhibiting protein synt hesis by binding to the L6 protein of the 30S ribosomal subunit. It is also effective against mycoplasma. Recommended 1 * 25 g
Katalogové číslo GENORC-175
Supplier:  G-Biosciences
Frequently Bought Together