CLINDAMYCIN HYDROCHLORIDE 1 * 200 mg
Catalog # EDQMC2250000
Supplier: EDQM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR®, Chlorite 1,000 mg/L in NaOH solution standard for ion chromatography
Supplier: VWR Chemicals
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets
Supplier: Mettler - Toledo
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
Polydisperse particle standards, CRM, Supelco®
Supplier: Merck
Magnesium standard for IC, TraceCERT®, Supelco®
Supplier: Merck
Buffer solutions
Supplier: REAGECON
Turbidity standards, Stablcal®
Supplier: Hach
pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
Supplier: Honeywell Chemicals
Lithium standard for IC, TraceCERT®, Supelco®
Supplier: Merck