MILK POWDER - ESSENTIAL NUTRIENTS AN 1 * 20 g
PROOERM-BD150
Frequently Bought Together
Accessories for Mettler balances
: Mettler - Toledo
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals