Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Potassium Hydroxide 85% pellets for analysis 1 * 1.000 g
Katalogové číslo APPC121515.1211
Dodavatel:
PanReac AppliChem
Certifikáty
Rozbalit obrázek
Potassium Hydroxide 85% pellets for analysis 1 * 1.000 g
Katalogové číslo APPC121515.1211
Dodavatel:
PanReac AppliChem
Certifikáty
Související informace
Dokumentace
Certifikáty
Hledat podle čísla šarže
Dokumentace
Frequently Bought Together
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Dodavatel: Merck
Haemalum (Mayer's), Sigma-Aldrich®
Dodavatel: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Dodavatel: VWR Chemicals
Visual colour kits, chloride, titrimetric with titration pipette, MQuant®, Supelco®
Dodavatel: Merck
Test kits, chlorine dioxide, Spectroquant®, Supelco®
Dodavatel: Merck
Test strips for peroxide, MQuant®, Supelco®
Dodavatel: Merck
1-Octanesulphonic acid sodium salt ≥98%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
pH indicator test strips, pH-Fix
Dodavatel: MACHEREY-NAGEL
Test strips, peroxide, Reflectoquant®, Supelco®
Dodavatel: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Dodavatel: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Dodavatel: Merck
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
Dodavatel: Merck
Test kits, photometric, ammonium, LCK cuvette tests
Dodavatel: Hach