Carazzi's Hematoxylin solution for clinical diagnosis 1 * 500 mL
Catalog # APPC255298.1610
Supplier: PanReac AppliChem
Carazzi's Hematoxylin solution for clinical diagnosis 1 * 500 mL
Catalog # APPC255298.1610
Supplier: PanReac AppliChem
Frequently Bought Together
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Supplier: Thermo Fisher Scientific