Tetrabutylammonium Hydroxide 0,1 mol/l (0,1N) in 2-propanol/methanol (11:1) volumetric solution 1 * 500 mL
Catalog # APPC183669.1610
Supplier: PanReac AppliChem
Tetrabutylammonium Hydroxide 0,1 mol/l (0,1N) in 2-propanol/methanol (11:1) volumetric solution 1 * 500 mL
Catalog # APPC183669.1610
Supplier: PanReac AppliChem
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Octane, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Chloroform 99.8% stabilised for analysis
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
1,4-Dioxane 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Butanone (Methylethylketone) pure 1 * 2,5 L
Supplier: PanReac AppliChem
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
1,3-Dioxolane 99.5+% stabilised, pure
Supplier: Thermo Fisher Scientific