ContactUsLinkComponent
Euro-norm plastic bin 40 litres, euro-standard 600 x 400 x 220 mm, with closed bottom and closed side-walls 1 * 1 items
Catalog # GORRKBE-183/01
Euro-norm plastic bin 40 litres, euro-standard 600 x 400 x 220 mm, with closed bottom and closed side-walls 1 * 1 items
Catalog # GORRKBE-183/01
Frequently Bought Together