ContactUsLinkComponent
Fridge, LKEXV 5000, capacity 554l, static cooling system, 5 glass shelve, solid doors, external thermostat , warning message, ambient temperature of +10 to 32 C, cooling from +1 to +15 C. 1 * 1 items
Catalog # LIEBLKEXV5000
Supplier:  Liebherr
Fridge, LKEXV 5000, capacity 554l, static cooling system, 5 glass shelve, solid doors, external thermostat , warning message, ambient temperature of +10 to 32 C, cooling from +1 to +15 C. 1 * 1 items
Catalog # LIEBLKEXV5000
Supplier:  Liebherr
Frequently Bought Together