ContactUsLinkComponent
Fridge, GG5260-974, capacity 513l, static cooling system, manual defrost ,housing stainless steel, slimline door, climatic class SN-ST, 220-240 V voltage. 1 * 1 items
Catalog # LIEBGG5260-974
Supplier:  Liebherr
Fridge, GG5260-974, capacity 513l, static cooling system, manual defrost ,housing stainless steel, slimline door, climatic class SN-ST, 220-240 V voltage. 1 * 1 items
Catalog # LIEBGG5260-974
Supplier:  Liebherr
Frequently Bought Together