FLUORESCENT DYE EU-STREPTAVIDIN FLUORESCENT DYE EU WITH STREPTAVIDIN CONJUGATE. FLUORESCENT DYE EU IS OPTIMIZED FOR TR-FRET APPLICA TIONS WITH LARGE STOKES SHIFTS AND EXTREMELY LONG EMISSION HALF-LIVES WHEN COMPARED TO TRADITIONAL FLUOROPHORES SUCH 1 * 100 µG
Catalog # ABNOU0310
Supplier: Abnova
Frequently Bought Together