ContactUsLinkComponent
FLUORESCENT DYE 405-I PHALLOIDIN FLUORESCENCE DYE 405-I WITH PHALLOIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENCE DY E 405-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10). FLUORESCENCE DYE 405-I LA 1 * 300 Assays
Catalog # ABNOU0278
Supplier:  Abnova
FLUORESCENT DYE 405-I PHALLOIDIN FLUORESCENCE DYE 405-I WITH PHALLOIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENCE DY E 405-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10). FLUORESCENCE DYE 405-I LA 1 * 300 Assays
Catalog # ABNOU0278
Supplier:  Abnova