ContactUsLinkComponent
Easy-fit torch, demountable, for volatile organics, with replaceable 0.8 mm id injector and base assembly, and replaceable outer and intermediate tubes, for radial viewing with Agilent 5100 Radial View/Vertical Dual View ICP-OES 1 * 1 items
Catalog # HEWLG8010-60235
Supplier:  Agilent
Easy-fit torch, demountable, for volatile organics, with replaceable 0.8 mm id injector and base assembly, and replaceable outer and intermediate tubes, for radial viewing with Agilent 5100 Radial View/Vertical Dual View ICP-OES 1 * 1 items
Catalog # HEWLG8010-60235
Supplier:  Agilent
Frequently Bought Together