ContactUsLinkComponent
Rabbit IgG polyclonal antibody for CD44 antigen(CD44) detection. Tested with WB, IHC-P, IHC-F, ICC in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTPA1021-2
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for CD44 antigen(CD44) detection. Tested with WB, IHC-P, IHC-F, ICC in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTPA1021-2
Supplier:  Boster Bio
Frequently Bought Together