ContactUsLinkComponent
COVERALL COLOR: WHITE SIZE: 2XL DESCRIPTION: ZIPPER IN FRONT, WITH HOOD, LATEX-FREE ELASTIC ON WRIST/ANKLE/WAIST MATERIAL: POLYPROPYLENE THICKNESS: 40 G/M PACKAGING: 1 PC/BAG, 50 PCS/CARTON 1 * 50 items
Catalog # GUNA140035
Supplier:  VWR Collection
COVERALL COLOR: WHITE SIZE: 2XL DESCRIPTION: ZIPPER IN FRONT, WITH HOOD, LATEX-FREE ELASTIC ON WRIST/ANKLE/WAIST MATERIAL: POLYPROPYLENE THICKNESS: 40 G/M PACKAGING: 1 PC/BAG, 50 PCS/CARTON 1 * 50 items
Catalog # GUNA140035
Supplier:  VWR Collection
Frequently Bought Together