ContactUsLinkComponent
Cylinder, capacity 1l, class A, graduation 10.0ml, stopper size 45/40, made of chemically resistant pyrex borosilicate glass, compli es with ISO 4788, supplied with polyethylene stopper, robust hexagonal glass foot. 1 * 10 items
Catalog # BIBB3212/16
Supplier:  SciLabware
undefined
Cylinder, capacity 1l, class A, graduation 10.0ml, stopper size 45/40, made of chemically resistant pyrex borosilicate glass, compli es with ISO 4788, supplied with polyethylene stopper, robust hexagonal glass foot. 1 * 10 items
Catalog # BIBB3212/16
Supplier:  SciLabware
Frequently Bought Together