ContactUsLinkComponent
Cylinder, capacity 250ml, class A, graduation 2.0ml, stopper size 29/32 ,made of chemically resistant pyrex borosilicate glass, complies with ISO4788, supplied with polyethylene stopper, robust hexagonal glass foot. 1 * 10 items
Catalog # BIBB3212/12
Supplier:  SciLabware
undefined
Cylinder, capacity 250ml, class A, graduation 2.0ml, stopper size 29/32 ,made of chemically resistant pyrex borosilicate glass, complies with ISO4788, supplied with polyethylene stopper, robust hexagonal glass foot. 1 * 10 items
Catalog # BIBB3212/12
Supplier:  SciLabware
Frequently Bought Together