ContactUsLinkComponent
Pocketmop in polysorb (textured polyester) 14” width. for use with qpsl-14 adapter. suitable for use in controlled environments and support areas. compatible with strong disinfectants, launderable, autoclavable. packaged in packs of 4, 6 packs to a c 1 * 24 items
Catalog # MICRPKM11-14IR
Supplier:  MICRONOVA
Pocketmop in polysorb (textured polyester) 14” width. for use with qpsl-14 adapter. suitable for use in controlled environments and support areas. compatible with strong disinfectants, launderable, autoclavable. packaged in packs of 4, 6 packs to a c 1 * 24 items
Catalog # MICRPKM11-14IR
Supplier:  MICRONOVA
Frequently Bought Together