ContactUsLinkComponent
Refrigerated Bath WCT80 for cold traps, capacity: 10 L, with Temperature Controller and digital Display, temperature range down to -80C, Bath: 220 x 220 x 210 mm, external dimension: 370 x 453 x 705 mm, refrigerator 736 W, Power supply: 230 V, 50/60 1 * 1 items
Catalog # WITGDH.WCT00180
Supplier:  witeg Labortechnik
Refrigerated Bath WCT80 for cold traps, capacity: 10 L, with Temperature Controller and digital Display, temperature range down to -80C, Bath: 220 x 220 x 210 mm, external dimension: 370 x 453 x 705 mm, refrigerator 736 W, Power supply: 230 V, 50/60 1 * 1 items
Catalog # WITGDH.WCT00180
Supplier:  witeg Labortechnik
Frequently Bought Together