ContactUsLinkComponent
Heating Mantles WHM, for 10 L reaction vessels w.bottom outlet, cylindrical, A-Ø 215 mm, Height 330 mm, without power controller, temperature range 450°C,1400 W, aluminium casing, power supply 230 V 1 * 1 items
Catalog # WITGDH.WHM12237
Supplier:  witeg Labortechnik
Heating Mantles WHM, for 10 L reaction vessels w.bottom outlet, cylindrical, A-Ø 215 mm, Height 330 mm, without power controller, temperature range 450°C,1400 W, aluminium casing, power supply 230 V 1 * 1 items
Catalog # WITGDH.WHM12237
Supplier:  witeg Labortechnik
Frequently Bought Together