ContactUsLinkComponent
WB SUBSTRATE KIT IV WB SUBSTRATE KIT IV IS A NON-ISOTOPIC, LUMINOL-BASED CHEMILUMINESCENCE SUBSTRATE, DESIGNED FOR THE CHEMILUMINES CENT DETECTION OF IMMOBILIZED PROTEINS AND IMMOBILIZED NUCLEIC ACIDS CONJUGATED WITH HORSERADISH PEROXIDASE 1 * 1 KIT
Catalog # ABNOKA4037
Supplier:  Abnova
WB SUBSTRATE KIT IV WB SUBSTRATE KIT IV IS A NON-ISOTOPIC, LUMINOL-BASED CHEMILUMINESCENCE SUBSTRATE, DESIGNED FOR THE CHEMILUMINES CENT DETECTION OF IMMOBILIZED PROTEINS AND IMMOBILIZED NUCLEIC ACIDS CONJUGATED WITH HORSERADISH PEROXIDASE 1 * 1 KIT
Catalog # ABNOKA4037
Supplier:  Abnova
Frequently Bought Together