ContactUsLinkComponent
Glass shaft, Pt junction, electrolyte KCl 3 mol/l, Silamid® reference, temp. sensor NTC 30 kOhm, sphere membrane, A glass, 1.5 m fix ed cable with digital plug, length 170 mm, 12 mm Ø, -5...+100 °C, 0...14 pH 1 * 1 items
Catalog # SCOI285100090
Supplier:  SI Analytics
Glass shaft, Pt junction, electrolyte KCl 3 mol/l, Silamid® reference, temp. sensor NTC 30 kOhm, sphere membrane, A glass, 1.5 m fix ed cable with digital plug, length 170 mm, 12 mm Ø, -5...+100 °C, 0...14 pH 1 * 1 items
Catalog # SCOI285100090
Supplier:  SI Analytics
Frequently Bought Together