ContactUsLinkComponent
Agilent J+W DB-Sim-Dist, 4 m, 0.25 mm, 0.5 µm, Low Thermal Mass (LTM) column module, 5 inch format, for Agilent Simulated Distillati on Analyzer 1 * 1 items
Catalog # HEWLG3900-65004
Supplier:  Agilent
Agilent J+W DB-Sim-Dist, 4 m, 0.25 mm, 0.5 µm, Low Thermal Mass (LTM) column module, 5 inch format, for Agilent Simulated Distillati on Analyzer 1 * 1 items
Catalog # HEWLG3900-65004
Supplier:  Agilent
Frequently Bought Together