ContactUsLinkComponent
Capillary kit, for Agilent Online-SPE Direct Injection Kit G4744A. Includes 3 capillaries, stainless steel, 0.12 x 280 mm, SL/SX fit tings (5067-4687), capillary, stainless steel, 0.25 x 250mm, S-S fittings (5067-5709), 2 blanking nuts, long, 10-32, 1 * 1 items
Catalog # HEWL5067-5710
Supplier:  Agilent
Capillary kit, for Agilent Online-SPE Direct Injection Kit G4744A. Includes 3 capillaries, stainless steel, 0.12 x 280 mm, SL/SX fit tings (5067-4687), capillary, stainless steel, 0.25 x 250mm, S-S fittings (5067-5709), 2 blanking nuts, long, 10-32, 1 * 1 items
Catalog # HEWL5067-5710
Supplier:  Agilent
Frequently Bought Together