EC HPLC column EC 125/4.6 NUCLEOSIL 100-10 NH2 length: 125 mm, ID: 4.6 mm pack of 1 1 * 1 items
Catalog # MANA720952.46
Supplier: MACHEREY-NAGEL
EC HPLC column EC 125/4.6 NUCLEOSIL 100-10 NH2 length: 125 mm, ID: 4.6 mm pack of 1 1 * 1 items
Catalog # MANA720952.46
Supplier: MACHEREY-NAGEL
Frequently Bought Together
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection