ContactUsLinkComponent
MolSieve 13X, 80/100, preconditioned, 41.28 mm coil, 10 feet (3.05 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Val ve Oven 1 * 1 items
Catalog # HEWLG3591-81203
Supplier:  Agilent
MolSieve 13X, 80/100, preconditioned, 41.28 mm coil, 10 feet (3.05 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Val ve Oven 1 * 1 items
Catalog # HEWLG3591-81203
Supplier:  Agilent
Frequently Bought Together