ContactUsLinkComponent
Splitter kit, 8 to 40 mL/min, preparative, for Agilent G4260B evaporative light scattering detector. Includes capillary to ELSD, 0.2 5 mm, 250 mm (5023-2263), capillary to fraction collector, 0.5 mm, 1,000 mm (5023-2261), tubing, PTFE, 0.7 mm id, 1.6 1 * 1 items
Catalog # HEWL5023-2257
Supplier:  Agilent
Splitter kit, 8 to 40 mL/min, preparative, for Agilent G4260B evaporative light scattering detector. Includes capillary to ELSD, 0.2 5 mm, 250 mm (5023-2263), capillary to fraction collector, 0.5 mm, 1,000 mm (5023-2261), tubing, PTFE, 0.7 mm id, 1.6 1 * 1 items
Catalog # HEWL5023-2257
Supplier:  Agilent
Frequently Bought Together