FABP4 inhibitor is a cell-permeable, potent inhibitor of FABP4 that targets the fatty acid-binding pocket (Ki < 2 nM).{25745} It is less effective against FABP3 and FABP5 (Kis = 250 and 350 nM, respectively).{25745} FABP4 inhibitor is orally active, 1 * 25 mg
Catalog # CAYM10010206-25
Supplier: Cayman Chemical
Frequently Bought Together