ContactUsLinkComponent
MreB perturbing compound A22 is a benzylisothiourea compound that interacts with the ATP binding site of MreB rapidly and reversibly .{25730} It blocks normal rod shape formation and inhibits chromosome partitioning in E. coli, inhibiting growth (MIC 1 * 10 mg
Catalog # CAYM15870-10
Supplier:  Cayman Chemical
MreB perturbing compound A22 is a benzylisothiourea compound that interacts with the ATP binding site of MreB rapidly and reversibly .{25730} It blocks normal rod shape formation and inhibits chromosome partitioning in E. coli, inhibiting growth (MIC 1 * 10 mg
Catalog # CAYM15870-10
Supplier:  Cayman Chemical
Frequently Bought Together