ContactUsLinkComponent
Cefazolin is a broad-spectrum, first-generation cephalosporin antibiotic that is active in vitro against most Gram-positive and Gram -negative bacteria.{25759} It inhibits bacterial cell wall synthesis, causing growth to cease without lysis (5 µg/ml 1 * 1 g
Catalog # CAYM15776-1
Supplier:  Cayman Chemical
Cefazolin is a broad-spectrum, first-generation cephalosporin antibiotic that is active in vitro against most Gram-positive and Gram -negative bacteria.{25759} It inhibits bacterial cell wall synthesis, causing growth to cease without lysis (5 µg/ml 1 * 1 g
Catalog # CAYM15776-1
Supplier:  Cayman Chemical
Frequently Bought Together