ContactUsLinkComponent
6-Thioguanine (6-TG) is a thio analog of the purine base guanine that incorporates into DNA during replication, inducing double-stra nd breaks that destabilize its structure and result in cytotoxicity.{25754,25756} Upon incorporation into DNA, 6-TG h 1 * 5 g
Catalog # CAYM15774-5
Supplier:  Cayman Chemical
6-Thioguanine (6-TG) is a thio analog of the purine base guanine that incorporates into DNA during replication, inducing double-stra nd breaks that destabilize its structure and result in cytotoxicity.{25754,25756} Upon incorporation into DNA, 6-TG h 1 * 5 g
Catalog # CAYM15774-5
Supplier:  Cayman Chemical
Frequently Bought Together