ContactUsLinkComponent
Yangonin is a natural kavalactone from the kava plant, P. methysticum. It enhances the binding of bicuculline at the γ-amino butyric acid (GABA) receptor GABAA at 1.0 µM.{25728} Yangonin also binds the central cannabinoid (CB1) receptor with a Ki va 1 * 5 mg
Catalog # CAYM14351-5
Supplier:  Cayman Chemical
Yangonin is a natural kavalactone from the kava plant, P. methysticum. It enhances the binding of bicuculline at the γ-amino butyric acid (GABA) receptor GABAA at 1.0 µM.{25728} Yangonin also binds the central cannabinoid (CB1) receptor with a Ki va 1 * 5 mg
Catalog # CAYM14351-5
Supplier:  Cayman Chemical
Frequently Bought Together