ContactUsLinkComponent
(-)-Epigallocatechin (EGC) is a major green tea polyphenol with antioxidant, anti-inflammatory, and anticancer activities. It has be en shown to scavenge DPPH radicals with an EC50 value of 0.01 mM and to prevent the growth of several different AML c 1 * 10 mg
Catalog # CAYM11809-10
Supplier:  Cayman Chemical
(-)-Epigallocatechin (EGC) is a major green tea polyphenol with antioxidant, anti-inflammatory, and anticancer activities. It has be en shown to scavenge DPPH radicals with an EC50 value of 0.01 mM and to prevent the growth of several different AML c 1 * 10 mg
Catalog # CAYM11809-10
Supplier:  Cayman Chemical
Frequently Bought Together