ContactUsLinkComponent
qPCR detection of Simkania negevensis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, En dogenous control, extraction control 1 * 150 Tests
Catalog # PATH-S.NEGEVEN
Supplier:  PRIMERDESIGN
qPCR detection of Simkania negevensis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, En dogenous control, extraction control 1 * 150 Tests
Catalog # PATH-S.NEGEVEN
Supplier:  PRIMERDESIGN
Frequently Bought Together