qPCR detection of Sus scrofa. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-S.SCROFA
Supplier: PRIMERDESIGN
Frequently Bought Together