ContactUsLinkComponent
qPCR detection of SARS coronavirus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endog enous control, extraction control 1 * 150 Tests
Catalog # PATH-SARS-COV
Supplier:  PRIMERDESIGN
qPCR detection of SARS coronavirus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endog enous control, extraction control 1 * 150 Tests
Catalog # PATH-SARS-COV
Supplier:  PRIMERDESIGN
Frequently Bought Together