ContactUsLinkComponent
qPCR detection of Bordetella pertussis. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome vi ruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-BPERT
Supplier:  PRIMERDESIGN
qPCR detection of Bordetella pertussis. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome vi ruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-BPERT
Supplier:  PRIMERDESIGN
Frequently Bought Together