ContactUsLinkComponent
qPCR detection of Burkholderia pseudomallei. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA geno me viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-B.PSE
Supplier:  PRIMERDESIGN
qPCR detection of Burkholderia pseudomallei. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA geno me viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-B.PSE
Supplier:  PRIMERDESIGN
Frequently Bought Together