ContactUsLinkComponent
qPCR detection of Campylobacter Coli. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viru ses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.COL
Supplier:  PRIMERDESIGN
qPCR detection of Campylobacter Coli. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viru ses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.COL
Supplier:  PRIMERDESIGN
Frequently Bought Together