ContactUsLinkComponent
qPCR detection of the active Diptheriae toxin domains A and B of Corynebacterium diphtheriae. Contains: Primer and probe mix, Revers e Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.DIP
Supplier:  PRIMERDESIGN
qPCR detection of the active Diptheriae toxin domains A and B of Corynebacterium diphtheriae. Contains: Primer and probe mix, Revers e Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.DIP
Supplier:  PRIMERDESIGN
Frequently Bought Together