ContactUsLinkComponent
qPCR detection of Influenza type A M1. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome vir uses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-FLUA1
Supplier:  PRIMERDESIGN
qPCR detection of Influenza type A M1. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome vir uses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-FLUA1
Supplier:  PRIMERDESIGN
Frequently Bought Together