qPCR detection of Trypanosoma cruzi. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-T.CRUZI
Supplier: PRIMERDESIGN
Frequently Bought Together