ContactUsLinkComponent
qPCR detection of Yellow Fever Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, End ogenous control, extraction control 1 * 150 Tests
Catalog # PATH-YFV
Supplier:  PRIMERDESIGN
qPCR detection of Yellow Fever Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, End ogenous control, extraction control 1 * 150 Tests
Catalog # PATH-YFV
Supplier:  PRIMERDESIGN
Frequently Bought Together